Anti-BAP1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1G8]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-60521APCEA 5290 DKK
NOVUNB110-60521APC
Anti-BAP1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1G8]
Antistoffer
Type: Primary
Antigen: BAP1 (BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase))
Clonality: monoclonal
Clone: 1G8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR