Anti-WT1 Mouse Monoclonal Antibody (DyLight® 650) [clone: 6F-H2]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-60011CEA 4980 DKK
NOVUNB110-60011C
Anti-WT1 Mouse Monoclonal Antibody (DyLight® 650) [clone: 6F-H2]
Antistoffer
Type: Primary
Antigen: WT1 (Wilms tumor 1)
Clonality: monoclonal
Clone: 6F-H2
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR