Anti-Capicua Rabbit Polyclonal Antibody (Biotin)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-59905BEA 4900 DKK
NOVUNB110-59905B
Anti-Capicua Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Capicua
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR