Anti-CD36 Mouse Monoclonal Antibody (DyLight® 650) [clone: D-2712]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-59724CEA 5360 DKK
NOVUNB110-59724C
Anti-CD36 Mouse Monoclonal Antibody (DyLight® 650) [clone: D-2712]
Antistoffer
Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: monoclonal
Clone: D-2712
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgA Kappa
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR