Anti-KIAA1524 Mouse Monoclonal Antibody (DyLight® 650) [clone: 2G10]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-59722CEA 5290 DKK
NOVUNB110-59722C
Anti-KIAA1524 Mouse Monoclonal Antibody (DyLight® 650) [clone: 2G10]
Antistoffer
Type: Primary
Antigen: KIAA1524 (KIAA1524)
Clonality: monoclonal
Clone: 2G10
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR