Anti-PMP22 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: Hu1]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-59086FEA 4580 DKK
NOVUNB110-59086F
Anti-PMP22 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: Hu1]
Antistoffer
Type: Primary
Antigen: PMP22 (peripheral myelin protein 22)
Clonality: monoclonal
Clone: Hu1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR