Anti-Spectrin beta 3 Rabbit Polyclonal Antibody (Biotin)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-58783BEA 5200 DKK
NOVUNB110-58783B
Anti-Spectrin beta 3 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Spectrin beta 3
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR