Anti-ROBO4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-58778HEA 5290 DKK
NOVUNB110-58778H
Anti-ROBO4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
The ROBO4 Antibody [HRP] from Novus Biologicals is a rabbit polyclonal antibody to ROBO4. This antibody reacts with human. The ROBO4 Antibody [HRP] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: ROBO4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR