Anti-ALOX5 Rabbit Polyclonal Antibody (DyLight® 488)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-58748GEA 4900 DKK
NOVUNB110-58748G
Anti-ALOX5 Rabbit Polyclonal Antibody (DyLight® 488)
Antistoffer
Type: Primary
Antigen: ALOX5 (arachidonate 5-lipoxygenase)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR