Anti-APOA5 Mouse Monoclonal Antibody (Biotin) [clone: 4H8H8E2]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-57307BEA 5290 DKK
NOVUNB110-57307B
Anti-APOA5 Mouse Monoclonal Antibody (Biotin) [clone: 4H8H8E2]
Antistoffer
Type: Primary
Antigen: APOA5 (apolipoprotein A-V)
Clonality: monoclonal
Clone: 4H8H8E2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR