Anti-APOA5 Mouse Monoclonal Antibody (DyLight® 550) [clone: 1G5G9]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-55454REA 4900 DKK
NOVUNB110-55454R
Anti-APOA5 Mouse Monoclonal Antibody (DyLight® 550) [clone: 1G5G9]
Antistoffer
Type: Primary
Antigen: APOA5 (apolipoprotein A-V)
Clonality: monoclonal
Clone: 1G5G9
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR