Anti-DNM1L Rabbit Polyclonal Antibody (DyLight® 650)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-55288CEA 5210 DKK
NOVUNB110-55288C
Anti-DNM1L Rabbit Polyclonal Antibody (DyLight® 650)
Antistoffer
Type: Primary
Antigen: DNM1L (dynamin 1-like)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR