Anti-SAT1 Rabbit Polyclonal Antibody (DyLight® 405)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-41622VEA 5300 DKK
NOVUNB110-41622V
Anti-SAT1 Rabbit Polyclonal Antibody (DyLight® 405)
Antistoffer
Type: Primary
Antigen: SAT1 (spermidine/spermine N1-acetyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR