Anti-ABHD5 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-41576EA 4160 DKK
NOVUNB110-41576 NOVUNB110-41576SS
Anti-ABHD5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ABHD5 (abhydrolase domain containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR