Anti-SLC2A1 Rabbit Polyclonal Antibody (Biotin)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB110-39113BEA 5900 DKK
NOVUNB110-39113B
Anti-SLC2A1 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: SLC2A1 (solute carrier family 2 (facilitated glucose transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR