Anti-CTGF Rabbit Polyclonal Antibody (DyLight® 550)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-724REA 5290 DKK
NOVUNB100-724R
Anti-CTGF Rabbit Polyclonal Antibody (DyLight® 550)
Antistoffer
Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR