Anti-VEGFA Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: VG1]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-664HEA 6340 DKK
NOVUNB100-664H
Anti-VEGFA Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: VG1]
Antistoffer
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone: VG1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR