Anti-rRNA Mouse Monoclonal Antibody (DyLight® 650) [clone: Y10b]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-662CEA 5900 DKK
NOVUNB100-662C
Anti-rRNA Mouse Monoclonal Antibody (DyLight® 650) [clone: Y10b]
Antistoffer
Type: Primary
Antigen: rRNA
Clonality: monoclonal
Clone: Y10b
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Pan
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR