Anti-6X His tag Mouse Monoclonal Antibody HRP (Horseradish Peroxidase) [clone: AD1.1.10]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-63173EA 5820 DKK
NOVUNB100-63173
Anti-6X His tag Mouse Monoclonal Antibody HRP (Horseradish Peroxidase) [clone: AD1.1.10]
Antistoffer
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone: AD1.1.10
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pan
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR