Anti-CAV1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7C8]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-615FEA 5640 DKK
NOVUNB100-615F
Anti-CAV1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7C8]
Antistoffer
Type: Primary
Antigen: CAV1 (caveolin 1, caveolae protein, 22kDa)
Clonality: monoclonal
Clone: 7C8
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR