Anti-BRCA1 Mouse Monoclonal Antibody (DyLight® 680) [clone: MU]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-600FREA 5290 DKK
NOVUNB100-600FR
Anti-BRCA1 Mouse Monoclonal Antibody (DyLight® 680) [clone: MU]
Antistoffer
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: MU
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR