Anti-BRCA1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MU]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-600APCEA 5290 DKK
NOVUNB100-600APC
Anti-BRCA1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MU]
Antistoffer
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: MU
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR