Anti-BRCA1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: RAY]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-598FEA 4900 DKK
NOVUNB100-598F
Anti-BRCA1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: RAY]
Antistoffer
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: RAY
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR