Anti-KMO Mouse Monoclonal Antibody (DyLight® 680) [clone: P3UI]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-597FREA 4900 DKK
NOVUNB100-597FR
Anti-KMO Mouse Monoclonal Antibody (DyLight® 680) [clone: P3UI]
Antistoffer
Type: Primary
Antigen: KMO (kynurenine 3-monooxygenase (kynurenine 3-hydroxylase))
Clonality: monoclonal
Clone: P3UI
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR