Anti-Langerin Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-56733EA 4510 DKK
NOVUNB100-56733
Anti-Langerin Rabbit Polyclonal Antibody
Antistoffer
Rabbit Polyclonal Langerin Antibody. Tested Applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin. Tested Reactivity: Mouse, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:500

Type: Primary
Antigen: Langerin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR