Anti-NIFK Mouse Monoclonal Antibody [clone: 1.8E149]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-56590EA 4380 DKK
NOVUNB100-56590
Anti-NIFK Mouse Monoclonal Antibody [clone: 1.8E149]
Antistoffer
Type: Primary
Antigen: NIFK (nucleolar protein interacting with the FHA domain of MKI67)
Clonality: monoclonal
Clone: 1.8E+149
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR