Anti-CHEK2 Mouse Monoclonal Antibody [clone: 73C175.1.1]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-56546EA 3270 DKK
NOVUNB100-56546
Anti-CHEK2 Mouse Monoclonal Antibody [clone: 73C175.1.1]
Antistoffer
Type: Primary
Antigen: CHEK2 (checkpoint kinase 2)
Clonality: monoclonal
Clone: 73C175.1.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR