Anti-MBD3 Mouse Monoclonal Antibody [clone: 106B691]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-56534EA 4090 DKK
NOVUNB100-56534
Anti-MBD3 Mouse Monoclonal Antibody [clone: 106B691]
Antistoffer
Type: Primary
Antigen: MBD3 (methyl-CpG binding domain protein 3)
Clonality: monoclonal
Clone: 106B691
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR