Anti-Caspase 8 Mouse Monoclonal Antibody [clone: 90A992]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-56527EA 4590 DKK
NOVUNB100-56527
Anti-Caspase 8 Mouse Monoclonal Antibody [clone: 90A992]
Antistoffer
The Caspase-8 Antibody (90A992) from Novus Biologicals is a mouse monoclonal antibody to Caspase-8. This antibody reacts with human, primate. The Caspase-8 Antibody (90A992) has been validated for the following applications: Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry-Paraffin, CyTOF-ready.

Simple Western , CyTOF-ready

Type: Primary
Antigen: Caspase 8
Clonality: Monoclonal
Clone: 90A992
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Primate
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR