Anti-RANKL Mouse Monoclonal Antibody [clone: 12A668]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-56512EA 4610 DKK
NOVUNB100-56512
Anti-RANKL Mouse Monoclonal Antibody [clone: 12A668]
Antistoffer
The TRANCE/TNFSF11/RANK L Antibody (12A668) from Novus Biologicals is a mouse monoclonal antibody to TRANCE / TNFSF11/RANK L. This antibody reacts with human, mouse, rat. The TRANCE / TNFSF11/RANK L Antibody (12A668) has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: RANKL
Clonality: Monoclonal
Clone: 12A668
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR