Anti-Corticotropin Releasing Factor Receptor 2 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-56485EA 4080 DKK
NOVUNB100-56485
Anti-Corticotropin Releasing Factor Receptor 2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Corticotropin Releasing Factor Receptor 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR