Anti-FLT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 4C11]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-527HEA 5180 DKK
NOVUNB100-527H
Anti-FLT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 4C11]
Antistoffer
Type: Primary
Antigen: FLT1 (fms-related tyrosine kinase 1)
Clonality: polyclonal
Clone: 4C11
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR