Anti-NOD2 Mouse Monoclonal Antibody (DyLight® 650) [clone: 2D9]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-524CEA 5310 DKK
NOVUNB100-524C
Anti-NOD2 Mouse Monoclonal Antibody (DyLight® 650) [clone: 2D9]
Antistoffer
Type: Primary
Antigen: NOD2 (nucleotide-binding oligomerization domain containing 2)
Clonality: monoclonal
Clone: 2D9
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR