Anti-PINK1 Rabbit Polyclonal Antibody (DyLight® 650)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-493CEA 4580 DKK
NOVUNB100-493C
Anti-PINK1 Rabbit Polyclonal Antibody (DyLight® 650)
Antistoffer
Type: Primary
Antigen: PINK1 (PTEN induced putative kinase 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR