Anti-DLX4 Rabbit Polyclonal Antibody (Biotin)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-481BEA 4580 DKK
NOVUNB100-481B
Anti-DLX4 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: DLX4 (distal-less homeobox 4)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR