Anti-Separase Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: XJ11-1B12]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-439HEA 5290 DKK
NOVUNB100-439H
Anti-Separase Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: XJ11-1B12]
Antistoffer
Type: Primary
Antigen: Separase
Clonality: monoclonal
Clone: XJ11-1B12
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR