Anti-RAD21 Mouse Monoclonal Antibody (Biotin) [clone: CM110-2C10]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-386BEA 4580 DKK
NOVUNB100-386B
Anti-RAD21 Mouse Monoclonal Antibody (Biotin) [clone: CM110-2C10]
Antistoffer
Type: Primary
Antigen: RAD21 (RAD21 homolog (S. pombe))
Clonality: monoclonal
Clone: CM110-2C10
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR