Anti-Cyr61/CCN1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-356HEA 5360 DKK
NOVUNB100-356H
Anti-Cyr61/CCN1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
The Cyr61/CCN1 Antibody [HRP] from Novus Biologicals is a rabbit polyclonal antibody to Cyr61/CCN1. This antibody reacts with human, rabbit. The Cyr61/CCN1 Antibody [HRP] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: Cyr61/CCN1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rabbit
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR