Anti-RPE65 Mouse Monoclonal Antibody (DyLight® 488) [clone: 401.8B11.3D9]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-355GEA 5370 DKK
NOVUNB100-355G
Anti-RPE65 Mouse Monoclonal Antibody (DyLight® 488) [clone: 401.8B11.3D9]
Antistoffer
Type: Primary
Antigen: RPE65 (retinal pigment epithelium-specific protein 65kDa)
Clonality: monoclonal
Clone: 401.8B11.3D9
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR