Anti-RNASEL Mouse Monoclonal Antibody (Biotin) [clone: 2E9]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-351BEA 4900 DKK
NOVUNB100-351B
Anti-RNASEL Mouse Monoclonal Antibody (Biotin) [clone: 2E9]
Antistoffer
Type: Primary
Antigen: RNASEL (ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent))
Clonality: monoclonal
Clone: 2000000000
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR