Anti-ATM Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10H11.E12]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-306FEA 4450 DKK
NOVUNB100-306F
Anti-ATM Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10H11.E12]
Antistoffer
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: monoclonal
Clone: 10H11.E12
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR