Anti-TP53BP1 Rabbit Polyclonal Antibody (Biotin)
Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NOVUNB100-305BEA 5520 DKK
NOVUNB100-305B
Anti-TP53BP1 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: TP53BP1 (tumor protein p53 binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse