Anti-KRT1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: LHK1]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-2756FEA 5290 DKK
NOVUNB100-2756F
Anti-KRT1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: LHK1]
Antistoffer
Type: Primary
Antigen: KRT1 (keratin 1)
Clonality: monoclonal
Clone: LHK1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR