Anti-LOX Rabbit Polyclonal Antibody (DyLight® 488)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-2530GEA 5060 DKK
NOVUNB100-2530G
Anti-LOX Rabbit Polyclonal Antibody (DyLight® 488)
Antistoffer
Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR