Anti-ABCA1 Mouse Monoclonal Antibody (Biotin) [clone: HJ1]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-2068BEA 4890 DKK
NOVUNB100-2068B
Anti-ABCA1 Mouse Monoclonal Antibody (Biotin) [clone: HJ1]
Antistoffer
Type: Primary
Antigen: ABCA1 (ATP-binding cassette, sub-family A (ABC1), member 1)
Clonality: monoclonal
Clone: HJ1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR