Anti-CANX Rabbit Polyclonal Antibody (DyLight® 405)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-1974VEA 4950 DKK
NOVUNB100-1974V
Anti-CANX Rabbit Polyclonal Antibody (DyLight® 405)
Antistoffer
Type: Primary
Antigen: CANX (calnexin)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR