Anti-SMN Mouse Monoclonal Antibody (DyLight® 405) [clone: 2B1]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-1936VEA 5500 DKK
NOVUNB100-1936V
Anti-SMN Mouse Monoclonal Antibody (DyLight® 405) [clone: 2B1]
Antistoffer
Type: Primary
Antigen: SMN
Clonality: monoclonal
Clone: 2B1
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR