Anti-MAPK8 Rabbit Polyclonal Antibody
Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NOVUNB100-1925SSEA 1750 DKK
NOVUNB100-1925SS NOVUNB100-1925
Anti-MAPK8 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MAPK8 (mitogen-activated protein kinase 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: C. elegans