Anti-CASR Mouse Monoclonal Antibody (DyLight® 405) [clone: HL 1499]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-1830VEA 4950 DKK
NOVUNB100-1830V
Anti-CASR Mouse Monoclonal Antibody (DyLight® 405) [clone: HL 1499]
Antistoffer
Type: Primary
Antigen: CASR (calcium-sensing receptor)
Clonality: monoclonal
Clone: HL 1499
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR