Anti-FANCD2 Rabbit Polyclonal Antibody (DyLight® 650)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-182CEA 5460 DKK
NOVUNB100-182C
Anti-FANCD2 Rabbit Polyclonal Antibody (DyLight® 650)
Antistoffer
Type: Primary
Antigen: FANCD2 (Fanconi anemia, complementation group D2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR